Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLK1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
KAM-CC105F KAM-CC105-D KAM-CC105-F KAM-CC105D
95045-922EA 547 USD InStock
CA95045-922 95045-922 CA95045-920 95045-920
Anti-PLK1 Mouse Monoclonal Antibody
Antibodies
The polo-like kinases (PLKs) function in cell-cycle progression and in multiple stages of mitosis. PLK1 is activated by phosphorylation of Thr210 at the G2/M phase boundary. Active PLK1 participates in mitotic entry through activation of Cdc25C and nuclear import of cyclin B1, thereby activating the Cdc2/cyclin B kinase.

  • For Western Blot

Recognizes human, mouse (weak) and rat Plk1. Detects a band of ~62kDa by Western blot.

Type: Primary
Antigen: PLK1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty