Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSK3A Mouse Monoclonal Antibody [clone: 1H8]

Supplier: Enzo Life Sciences
KAM-ST002F KAM-ST002-D KAM-ST002-F KAM-ST002D
95045-934EA 552 USD InStock
95045-934 CA95045-932 95045-932 CA95045-934
Anti-GSK3A Mouse Monoclonal Antibody [clone: 1H8]
Antibodies
Glycogen Synthase Kinase 3β (GSK-3β) is a unique serine/threonine kinase that is inactivated by phosphorylation. In response to insulin binding, PKB/AKT phosphorylates GSK-3β on serine 9, which prevents GSK-3β from phosphorylating glycogen synthase. Unphosphorylated glycogen synthase is active and able to synthesize glycogen. GSK-3β is also unique in that it requires a substrate that has been phosphorylated by a distinct kinase before it can phosphorylate the substrate. This phosphate priming mechanism explains why phosphorylation of serine 9 inactivates GSK-3β. The phosphorylated serine binds to the GSK-3β priming phosphate position and prevents binding of alternative substrates. In addition to insulin signaling, GSK-3β participates in the Wnt signaling pathway, where it forms a complex with axin, beta-catenin and adenomatous polyposis coli (APC) protein. In the presence of Wnts, GSK-3β is unable to phosphorylate beta-catenin, which leads to stabilization of beta-catenin. The Wnt pathway inactivates GSK-3β via the proteins, Dishevelled and FRAT, which disrupt the interaction of GSK-3β with axin, beta-catenin, and APC. Clinically, there is considerable interest in GSK-3β inhibitors because they may mimic the effect of insulin or reduce the hyperphosphorylation of Tau that is observed in Alzheimer's Disease.

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, hamaster, monkey, pig, rabbit and sheep GSK-3alpha/beta. Detects bands of ~51 and ~47kDa by Western blot.

Type: Primary
Antigen: GSK-3ALPHA
Clonality: Monoclonal
Clone: 1H8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Sheep (Ovine), Dog (Canine), Pig (Pig, Swine), Monkey, Cow (Bovine), Hamaster, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty