Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKA Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAS-PK017-D KAS-PK017D KAS-PK017-F KAS-PK017F
95046-002EA 217 USD InStock
95046-002 CA95046-002 95046-004 CA95046-004
Anti-PKA Rabbit Polyclonal Antibody
Antibodies
PKA (Protein Kinase A), also known as cAMP-dependent protein kinase, phosphorylates serine or threonine residues in target protein in response to elevated levels of cAMP. Inactive PKA exists as a tetrameric protein composed of two regulatory (R) subunits and two catalytic (C) subunits. Activation occurs when two cAMP molecules bind to each R subunit which causes a conformational change that releases the active C subunits.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus PKA. Detects a band of ~40kDa by Western blot.

Type: Primary
Antigen: PKA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty