Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Enzo Life Sciences
SAB-600PE-D SAB-600PE-F SAB-600PEF SAB-600PED
95046-074EA 280 USD InStock
95046-074 95046-076 CA95046-074 CA95046-076
Anti-IgG1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Isotype matched controls are useful in flow cytometry to determine a gross estimate of non-specific binding that may be observed with a specific antibody of interest. In general, an isotype control should be of the same species and isotype as the antibody of interest, used at the same concentration, and conjugated to the same fluorophore. This reagent is an IgG1 isotype mouse monoclonal antibody (Clone # MOPC-21) conjugated to phycoerythrin (PE). It is intended for use as an isotype control antibody for flow cytometry.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty