Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SOD2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
SOD-110F ADI-SOD-110-F SOD-110D ADI-SOD-110-D
95046-102EA 265 USD InStock
CA95046-104 95046-102 95046-104 CA95046-102
Anti-SOD2 Rabbit Polyclonal Antibody
Antibodies
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Mn SOD. Detects a band of ~25kDa by Western blot.

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty