Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP47 Mouse Monoclonal Antibody [clone: M16.10A1]

Supplier: Enzo Life Sciences
SPA-470J ADI-SPA-470-J
95046-132EA 2350 USD InStock
95046-132 CA95046-132
Anti-HSP47 Mouse Monoclonal Antibody [clone: M16.10A1]
Antibodies
Type: Primary
Antigen: HSP47
Clonality: Monoclonal
Clone: M16.10A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty