Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTGES3 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-670-D ADI-SPA-670-F SPA-670D SPA-670F
95046-152EA 293 USD InStock
95046-152 95046-154 CA95046-152 CA95046-154
Anti-PTGES3 Rabbit Polyclonal Antibody
Antibodies
p23 is a ubiquitous, highly conserved co-chaperone for Hsp90 that participates in the folding of a number of cell regulatory proteins including progesterone and glucocorticoid receptors, HSF1, and telomerase. p23 is thought to modulate Hsp90 activity in the last stages of the chaperoning pathway by binding and stabilizing Hsp90 in its ATP-bound state.

  • For Western Blot

Recognizes human, mouse and rat p23. Detects a band of ~23kDa by Western blot.

Type: Primary
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty