Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90A Rat Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Enzo Life Sciences
SPA-840HRPD SPA-840HRP-F SPA-840HRP-D SPA-840HRPF
95046-260EA 634 USD InStock
CA95046-258 95046-260 CA95046-260 95046-258
Anti-HSP90A Rat Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The 90 kDa molecular chaperone family includes 90 kDa heat shock protein Hsp90 and 94 kDa glucose-regulated protein grp94, both major molecular chaperones of the cytosol and the endoplasmic reticulum. Mammalian cells contain isoforms Hsp90 alpha and Hsp90 beta, encoded by separate genes. The amino acid sequences of human and yeast Hsp90 alpha are 85% and 90% homologous to those of Hsp90 beta, respectively. All known members of the Hsp90 protein family are highly conserved, especially in the N -terminal and C-terminal regions containing independent chaperone sites with different substrate specificity. These ubiquitous and highly conserved proteins account for 1-2% of all cellular proteins in most cells. Hsp90 functions as part of the cell

  • For Western Blot

Recognizes human and chicken HSP90alpha. Detects a band of ~90kDa by Western blot.

Type: Primary
Antigen: HSP90A
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty