Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KDEL Receptor Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAA-PT048D VAA-PT048-F VAA-PT048-D VAA-PT048F
95046-432EA 280 USD InStock
CA95046-434 95046-432 CA95046-432 95046-434
Anti-KDEL Receptor Mouse Monoclonal Antibody
Antibodies
The tetrapeptide KDEL, located at the carboxy-terminal sequences of luminal proteins, is a retrieval motif essential for the precise sorting of these proteins along the secretory pathway. KDEL proteins perform essential functions in the endoplasmic reticulum (ER) related to protein folding as well as assembly. The localization of chaperones and other soluble proteins to the ER is achieved by their continuous retrieval from post-ER compartments by the KDEL receptor (Erd2p), which is a membrane protein localized in the Golgi apparatus.

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rat KDEL Receptor. Detects a band of ~23kDa by Western blot.

Type: Primary
Antigen: KDEL Receptor
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty