Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SAP97 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-PS005-D VAM-PS005D VAM-PS005-F VAM-PS005F
95046-442EA 265 USD InStock
95046-442 CA95046-442 95046-444 CA95046-444
Anti-SAP97 Mouse Monoclonal Antibody
Antibodies
Synapse-associated proteins SAP97 and SAP102 display extensive sequence homology to the PSD 95 family of proteins that facilitate ion channel clustering at synaptic terminals. In addition to the CNS, SAP97 is also detected at the basal lateral membrane between a variety of epithelial cells. SAP102 associates with NMDA receptors through the C-terminal NR2B subunit of the receptor. The N-terminal SAP97 PDZ domain mediates its interaction with the synaptic ras-GTPase activating protein SynGAP.

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot, EM

Recognizes human, mouse, rat, and dog SAP97. Detects bands of ~97kDa and ~140kDa by Western blot.

Type: Primary
Antigen: SAP97
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty