Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GS28 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VAM-PT047-F VAM-PT047F VAM-PT047D VAM-PT047-D
95046-448EA 662 USD InStock
95046-448 CA95046-446 CA95046-448 95046-446
Anti-GS28 Mouse Monoclonal Antibody
Antibodies
Golgi SNARE of 28 kDa (GS28), also known as p28 or GOS28, is a 28 kDa integral membrane protein on the surface of the Golgi apparatus that serves as a t-SNARE in ER to Golgi transport. The amino-terminal of GS28 is exposed to the cytosol and is anchored to the cis Golgi via a 20 amino acid carboxyl-terminal hydrophobic tail. GS28 co-immunoprecipitates complexes consisting of Syntaxin 5, Rbet1, Membrin, Rsec22, and Rsly1, and is therefore implicated in ER to-Golgi or intra-Golgi vesicle transport.

  • For Western Blot, EM

Recognizes mouse, rat, bovine, dog, chicken, hamster, pig, rabbit and sheep GS28. Detects a band of ~28kDa by Western blot.

Type: Primary
Antigen: GS28
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty