Would you like to log in again?
Stop Solution 2, Enzo Life Sciences
Supplier: Enzo Life Sciences
80-1804 ADI-80-1804 ADI-80-0377 80-0377
95052-788EA 48 USD InStock
CA95052-788 95052-788 95045-200 CA95045-200
Stop Solution 2, Enzo Life Sciences
Assays ELISAs
Acid stop solution (1 N HCl) intended for use as an assay stop reagent for colorimetric HRP/TMB reactions.
Each 50 ml bottle is sufficient for use with 5×96 wells microtiter plates.
Ordering information: Additional assay development reagents are available as companion products to IMMUNOSET® products.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty