Would you like to log in again?
HyClone™ AdvanceSTEM, HEPES Buffer, ES Qualified, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
SH30851.01 SH30851.01
95053-212EA 76.26 USD InStock
CA95053-212 95053-212
HyClone™ AdvanceSTEM, HEPES Buffer, ES Qualified, HyClone products (Cytiva)
Buffers Cell Culture Buffers
HyClone AdvanceSTEM ES Qualified HEPES is better at maintaining physiological pH despite changes in carbon dioxide as compared to bicarbonate buffers. Providing optimal culture conditions can be one of the greatest challenges in stem cell research, particularly in keeping cells in an undifferentiated state or directing differentiation when desired. HyClone AdvanceSTEM line of stem cell culture products can help you meet those challenges.
  • ES cell-qualified with HyClone's ES Screened FBS — no prescreening required.
  • Mycoplasma-free and sterility tested
  • pH range: 7.2 to 7.5
  • 0.1 µm sterile filtered
Providing optimal culture conditions can be one of the greatest challenges in stem cell research, particularly in keeping cells in an undifferentiated state or directing differentiation when desired. HyClone AdvanceSTEM line of stem cell culture products can help you meet those challenges.
Classic culture conditions for 129-derived murine embryonic stem cells (mESCs) include culturing in medium containing DMEM, FBS, LIF and co-culturing with primary mouse embryonic fibroblasts (MEFs). Pluripotent mESCs have a very distinct morphology when cultured under these conditions; growing as tightly clustered colonies with smooth phase bright borders. When co-cultured in ES cell media with a mouse embryonic fibroblast (MEF) feeder layer, these stem cell cultures are suitable for electroporation, expansion, or further experimentation.
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty