Would you like to log in again?
HyClone™ AdvanceSTEM™ Cryopreservation Media, Cytiva
Supplier: HyClone products (Cytiva)
HyClone™
SH30894.01 SH30894.01
95053-252EA 191.89 USD InStock
CA95053-252 95053-252
HyClone™ AdvanceSTEM™ Cryopreservation Media, Cytiva
Cell Freezing Media
HyClone AdvanceSTEM Cryopreservation Medium is formulated to support the cryopreservation of a wide range of cell types. Formulated to support the cryopreservation of a variety of cell types, including Human Mesenchymal Stem Cells, multipotent unrestricted stem cells of Umbilical Cord Blood and Human Amniotic Epithelial Stem Cells.
Formulated to support the cryopreservation of a variety of cell types, including Human Mesenchymal Stem Cells, multipotent unrestricted stem cells of Umbilical Cord Blood and Human Amniotic Epithelial Stem Cells.
Caution: Storage < -10° C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty