Would you like to log in again?
Trypsin Protease, HyClone™
Supplier: HyClone products (Cytiva)
SV30031.01 SH30042.01 SH30236.01 SV30037.01 SV30031.01 SH30236.01 SH30042.02 SH30236.02 SH30042.02 SH30042.01 SV30037.01 SH30236.02
16777-166EA 20.5 USD InStock
16777-166 95053-258 CA82003-690 82003-690 CA95053-258 CA95053-260 82003-688 95053-260 CA16777-202 CA16777-166 16777-202 CA82003-688
Trypsin Protease, HyClone™
Cell Culture Supplements and Additives
Dissociation reagents, such as trypsin, are used to detach and disaggregate adherent cells from the surface or substrate to which they are attached, as well as for dissociating tissues for primary cell culture.
  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty