Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Exendin-4 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ABS-012-24-1
95053-344EA 4190 USD InStock
95053-344
Anti-Exendin-4 Mouse Monoclonal Antibody
Antibodies
Exendin-4 is a 39-amino-acid peptide found in venom from the Gila monster Heloderma suspectum. It is a member of the glucagon-secretin family of peptide hormones and neuropeptides. Exendin-4 is a potent agonist of the GLP-1 receptor and hence a potent stimulator of insulin secretion.

  • For ELISA, Flow Cytometry

Recognizes human exendin-4. Detects a band of ~4kDa by Western blot.

Type: Primary
Antigen: EXENDIN-4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 ?
Reactivity: Heloderma Suspectum

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty