Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Complement C5B-9 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
DIA-011-01-1
95053-720EA 3760 USD InStock
95053-720
Anti-Complement C5B-9 Mouse Monoclonal Antibody
Antibodies
C5b-9, also known as the terminal complement complex (TCC), consists of C5b, C6, C7, C8 and C9, and it forms the membrane attack complex (MAC) as well as the non-lytic fluid-phase SC5b-9 complex (with protein S). The MAC forms channels in target cell membranes leading to cell lysis by osmotic leakage. The complexes contain neoantigens absent from the individual native components from which they are formed, and the antibody is directed against a neoepitope exposed on C9 when incorporated into the TCC.

  • For ELISA, Flow Cytometry, Immunohistochemistry

Recognizes human complement component C5b-9.

Type: Primary
Antigen: COMPLEMENT C5B-9
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a ?
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty