Would you like to log in again?
NGAL (Human) ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BPD-KIT-036
95054-124EA 1000 USD InStock
95054-124
NGAL (Human) ELISA Kit, Enzo Life Sciences
Assays ELISAs
A colorimetric immunometric ELISA kit for the measurement of NGAL in human samples
  • Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
  • Highly sensitive measurement, detecting as low as 4 pg/mL of NGAL
  • Ready-to-use pre-coated plate and individual calibrators save time and reduce errors
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
For the quantitative determination of NGAL in human urine, plasma, serum, tissue extracts, and culture supernatants.
Absorbance is read at 450 nm.
Delivery information: Kit consists of 12 strips of 8 microwells each coated with anti-NGAL (96 wells) + frame, sample diluent, NGAL calibrators 1-8; 0, 10, 25, 50, 100, 250, 500, 1000 pg/mL, 25X wash solution concentrate, biotinylated NGAL antibody, HRP-Streptavidin conjugate, TMB substrate and stop solution.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty