Would you like to log in again?
Blood Pressure Cuff Barriers, Single-Use, TIDI®
Supplier: TIDI
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
916113 916114 916115
95055-000CS 89.9 USD InStock
95055-000 95054-998 95055-002
Blood Pressure Cuff Barriers, Single-Use, TIDI®
First Aid Supplies Surgical Drapes
These barriers are made of soft Fabricel® tissue fused with leakproof polyethylene film that provides 99.9% protection against bacteria pass-through.
The barriers can be applied to arms or legs in seconds and will not alter blood pressure readings. Available in three sizes to fit any size blood pressure cuff.
Packaging: Packaged in dispenser boxes of 50.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty