Would you like to log in again?
RESOURCE S Columns, Cytiva
Supplier: Cytiva
17-1178-01 17-1178-01 17-1180-01 17-1180-01
95056-190EA 4148.51 USD InStock
CA95056-190L CA95055-802L 95056-190 95055-802
RESOURCE S Columns, Cytiva
Chromatography Columns
RESOURCE™ S columns are prepacked with SOURCE 15S, a strong cation exchanger for high-resolution polishing purification of proteins, at high flow rates.
  • Sulfoethyl (S) strong cation exchanger for high resolution ion exchange chromatography
  • Mono-sized 15 µm rigid beads give low back pressure and high flow rates allowing for high productivity
  • High chemical stability enables wide range of working conditions with good resistance to cleaning conditions at high pH
Rigid, monodispersed spherical SOURCE 15S resin beads (15 µm) with controlled pore-size distribution facilitate high-resolution purification at high flow rates. Hydrophilization of the beads further minimizes nonspecific adsorption and allows for high recovery rates for purified samples.
The material of the RESOURCE™ S column body is PEEK. This ion exchange chromatography column generates low back pressures and high performance, making it excellent for ion exchange (IEX) chromatography on a large scale.
Delivery information: Includes connectors for high- and low-pressure chromatography equipment and instructions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty