Would you like to log in again?
HiTrap Columns, Phenyl HP, Cytiva
Supplier: Cytiva
HiTrap™
17-5195-01 17-1351-01 17-1351-01 17-5195-01
95055-812PK 315.06 USD InStock
CA95055-812L 95055-812 95056-122 CA95056-122L
HiTrap Columns, Phenyl HP, Cytiva
Chromatography Columns
HiTrap Phenyl HP are prepacked Phenyl Sepharose High Performance columns for high-resolution hydrophobic interaction chromatography (HIC).
  • Provide convenient and fast small-scale protein purifications
  • Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
  • High flow rates and high capacities
  • Excellent and easy scale-up achieved by connecting several HiTrap columns in series or using HiPrep columns
Phenyl Sepharose High Performance is a well-established aromatic HIC resin, designed for intermediate and polishing steps where high resolution has priority.
The columns are used for small-scale protein purification as well as for screening of binding and elutions conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty