Would you like to log in again?
Ni Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-5268-02 17-5268-02 17-5268-01 17-5268-01
95055-838EA 514.25 USD InStock
95055-838 CA95055-838L 95056-210 CA95056-210L
Ni Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Ni Ni Sepharose High Performance affinity media for high-resolution, high capacity purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
  • High-performance purification of histidine-tagged proteins
  • Negligible leakage of Ni₂₊ ions
  • High binding capacity, at least 40 mg/ml medium
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives
  • 34 µm bead size for improved resolution
Ni Sepharose High Performance consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with Ni₂₊, which will selectively retain proteins with exposed histidine groups. The high stability and broad compatibility of Ni Sepharose High Performance maintains biological activity and increases product yield, at the same time as it greatly expands the range of suitable operating conditions. Chelating agents such as EDTA, EGTA , citrate should not be used.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty