Would you like to log in again?
Superose™ 12 Gel Filtration Media, Prep Grade
Supplier: Cytiva
Superose™
17-0536-01 17-0536-01
95055-854EA 894.81 USD InStock
CA95055-854L 95055-854
Superose™ 12 Gel Filtration Media, Prep Grade
Protein Purification Protein Purification Resins
Good resolution separations of proteins and other biomolecules according to size, Wide separation range – separations in two molecular weight ranges from 1,000 up to 5 million, Excellent reproducibility and durability
  • Excellent reproducibility and durability
Achieve high-resolution separations across exceptionally broad molecular-weight ranges. Low ionic strength eluents (< 0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds. High performance gel filtration media specially designed for preparative purification of biomolecules. Superose 6 is designed for high-resolution chromatography. Superose 6 prep grade permits easy, efficient scale-up to preparative separations. Superose 12 is for high resolution purification of proteins from 1 × 103 to 3 × 105 molecular weight, while Superose 12 prep grade is useful for preparative purification of these higher molecular weight proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty