Would you like to log in again?
HiTrap IEX Selection Kit, Cytiva
Supplier: Cytiva
HiTrap™
17-6002-33 17-6002-33
95055-856EA 394.43 USD InStock
CA95055-856L 95055-856
HiTrap IEX Selection Kit, Cytiva
Chromatography Columns
HiTrap IEX Selection Kit is used for screening of the optimal Cytiva ion exchange resin for specific application and development work.
  • Fast and easy selection: Seven prepacked HiTrap columns, each containing a different Sepharose ion exchange resin (medium)
  • Convenient way to screen for the optimal Cytiva anion exchange (aex) or cation exchange (cex) resin for a given application
  • Excellent for optimization studies; buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
  • HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
Cytiva's HiTrap IEX Selection Kit contains a variety of anion and cation exchange resins, allowing users to screen for the most appropriate column.
Ordering information: Prepacked HiTrap IEX columns are also available individually.
Delivery information: The kit includes seven 1 ml HiTrap columns packed with the following ion exchange resins: SP Sepharose Fast Flow cex column (strong cation exchanger), SP Sepharose XL cex column (strong cation exchanger), CM Sepharose Fast Flow cex column (weak cation exchanger), Q Sepharose Fast Flow aex column (strong anion exchanger), Q Sepharose XL aex column (strong anion exchanger), DEAE Sepharose Fast Flow aex column (weak anion exchanger), ANX Sepharose 4 Fast Flow aex column (weak anion exchanger).
Packaging: The ion exchange resins in the columns are offered as bulk resin and in other prepacked formats.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty