Would you like to log in again?
Spin Adapters for Gravity Columns, Cytiva
Supplier: Cytiva
28-9232-44 28-9232-43 28-9232-45 28-9232-44 28-9232-45 28-9232-43
95056-064PK 27.72 USD InStock
95056-064 CA95055-862L CA95056-064L 95055-862 95056-066 CA95056-066L
Spin Adapters for Gravity Columns, Cytiva
Chromatography Columns
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel, High desalting capacity, The use of centrifugation protocol with all gravity columns.
  • Convenient adapters, enable spinning of all gravity columns
Adapters are polypropylene rings designed for use with the gravity columns PD MiniTrap G-25, PD MidiTrap G-25, and PD-10 Desalting Columns when the spin protocol is used. MiniSpin Adapter fits PD MiniTrap G-25 columns, MidiSpin Adapter fits PD MidiTrap G-25 columns, and PD-10 Spin Adapter fits PD-10 Desalting Columns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty