Would you like to log in again?
HiTrap Columns, rProtein A FF, Cytiva
Supplier: Cytiva
HiTrap™
17-5079-02 17-5080-01 17-5080-01 17-5080-02 17-5080-02 17-5079-01 17-5079-01 17-5079-02
95056-008EA 1003.95 USD InStock
95056-008 CA95056-090L CA95055-868L 95055-868 CA95056-008L 95056-090 CA95056-088L 95056-088
HiTrap Columns, rProtein A FF, Cytiva
Chromatography Columns
HiTrap rProtein A FF columns are prepacked with rProtein A Sepharose Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources, including cell supernatants and ascites.
  • Quick, convenient preparative purification of monoclonal antibodies
  • Well-established protein A-based resin used in many approved MAb processes
  • Simple operations with a syringe, pump, or high-performance chromatography system, such as ÄKTA design
Recombinant protein A exhibits similar Fc region specificity to that of native protein A but shows enhanced binding capacity and fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
The recombinant protein A ligand (produced in E. coli) is coupled to Sepharose Fast Flow by a technique which generates a stable thioether linkage between rProtein A and the base matrix. The coupling technique is optimized to give high binding capacity for IgG.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty