Would you like to log in again?
HiTrap Columns, IgM Purification HP, Cytiva
Supplier: Cytiva
HiTrap™
17-5110-01 17-5110-01
95055-888PK 870.77 USD InStock
CA95055-888L 95055-888
HiTrap Columns, IgM Purification HP, Cytiva
Chromatography Columns
HiTrap IgM Purification HP is a 1 ml column prepacked with Sepharose High Performance for fast and efficient purification of monoclonal IgM.
  • Optimized for fast, efficient purification of monoclonal IgM from hybridoma cell culture
  • Prepacked with a thiophilic adsorption resin, 2-mercaptopyridine coupled to Sepharose High Performance
  • Recovery of IgM is high with excellent activity retention
  • The top and bottom frits are manufactured from porous polyethylene
Thiophilic adsorption is promoted by water-structuring salts. It has been suggested that the interaction of IgM and ligand results from combined electron donating and accepting action of the ligand or, alternatively, a mixed mode hydrophilic-hydrophobic interaction.
Delivery information: Columns are delivered with a stopper on the inlet and a snap-off end on the outlet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty