Would you like to log in again?
HiTrap Columns, IgY Purification HP, Cytiva
Supplier: Cytiva
HiTrap™
17-5111-01 17-5111-01
95055-890EA 853.72 USD InStock
95055-890 CA95055-890L
HiTrap Columns, IgY Purification HP, Cytiva
Chromatography Columns
HiTrap IgY Purification HP is a 5 ml column prepacked with Sepharose High Performance for fast and easy purification of IgY from egg yolk.
  • Each column binds IgY from a one-quarter portion of egg yolk (approx. 100 mg IgY)
  • For greater capacity, several columns may be connected in series
  • Packed with a thiophilic adsorption medium, the ligand, 2-mercaptopyridine is coupled to Sepharose High Performance
  • Recovery and purity of IgY are excellent
  • Can be operated with a liquid chromatography system such as ÄKTA design
  • The top and bottom frits are manufactured from porous polyethylene
HiTrap IgY Purification HP is packed with a thiophilic adsorption medium, 2-mercaptopyridine coupled to Sepharose High Performance. Thiophilic adsorption is promoted by water-structuring salts. It has been suggested that the interaction of IgY and ligand results from combined electron donating and accepting action of the ligand or, alternatively, a mixed mode hydrophilic-hydrophobic interaction.
Delivery information: Connectors for usage with different chromatography systems and other equipment are included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty