Would you like to log in again?
HiTrap Columns, Streptavidin HP, Cytiva
Supplier: Cytiva
HiTrap™
17-5112-01 17-5112-01
95055-892PK 1089.72 USD InStock
CA95055-892L 95055-892
HiTrap Columns, Streptavidin HP, Cytiva
Chromatography Columns
HiTrap Streptavidin HP is a column prepacked with Streptavidin Sepharose High Performance for purification of biotinylated proteins and biomolecules.
  • Convenient, fast, reliable purification of biotinylated compounds
  • The strong interactions of biotin and streptavidin, as well as the somewhat weaker interactions of 2-iminobiotin and streptavidin are utilised
  • The column can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System
Streptavidin Sepharose™ High Performance is also available in lab packs, using the same purified streptavidin isolated from Streptomyces avidinii bound to Sepharose™ High Performance. The prepacked medium ensures reproducibility, high performance, and excellent physical and chemical stability.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty