Would you like to log in again?
SOURCE™ 15S PE Columns, Cytiva
Supplier: Cytiva
Source™
17-5182-01 17-5182-01
95055-902EA 1645.46 USD InStock
95055-902 CA95055-902L
SOURCE™ 15S PE Columns, Cytiva
Chromatography Columns
Source 15S 4.6/100 PE is a strong cation exchanger prepacked in a Tricorn column for intermediate lab-scale purification and large-scale polishing of biomolecules with ion exchange chromatography.
  • SOURCE 15S resins are an excellent choice for fast, high-resolution purifications and easy scale-up
  • Rigid, monodispersed, spherical particles with controlled pore-size distribution offer excellent flow characteristics, sample loading of up to 25 mg of protein/ml, and improved stability in organic solvents and pH extremes
  • Improved capacity compared to MonoBeads but with a slightly lower resolution
The small (15 µm) monodisperse bead give high-resolution purification at high flow rates. In addition, hydrophilization of the beads minimizes nonspecific adsorption and allows high recovery of purified sample. The material of the column body is PEEK (polyetheretherketone).
SOURCE 30Q and 30S resins with a 30 µm bead size are designed for intermediate purification and large-scale polishing and allow higher flow rates.
Delivery information: Includes Valco fittings for ÄKTAdesign systems and are supplied with M6 connectors for FPLC System.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty