Would you like to log in again?
Dextrin Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
28-9355-97 28-9355-97 28-9355-98 28-9355-98
95055-918EA 4032.94 USD InStock
95055-918 95055-916 CA95055-916L CA95055-918L
Dextrin Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Dextrin Sepharose™ High Performance is an affinity media for purification of recombinant proteins tagged with Maltose Binding Protein (MBP).
  • The small, evenly sized beads ensure that MBP-tagged proteins elute in narrow peaks, thus minimizing the need for further concentration steps
  • Physiological conditions and mild elution preserve target protein activity
  • Dextrin Sepharose High Performance tolerates all commonly used aqueous buffers and is easily regenerated using 0.5 M NaOH allowing the same column to be used for repeated purifications
Dextrin Sepharose High Perfromance is an affinity media for purification of proteins tagged with Maltose Binding Protein (MBP). Dextrin Sepharose High Performance and provide convenient affinity purification of recombinant proteins tagged with maltose binding protein (MBP). This robust and stable Sepharose High Performance based medium enables elution of target protein in narrow peaks, which minimizes the need for further concentration steps.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty