Would you like to log in again?
QAE Sephadex™ Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Sephadex™
17-0190-01 17-0190-01 17-0200-01 17-0200-01
95055-934EA 562.01 USD InStock
CA95055-934L CA95055-932L 95055-934 95055-932
QAE Sephadex™ Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix.
  • Strong anion exchanger suitable batch techniques
  • High binding capacities
  • Stable to all commonly used aqueous buffers additives such as 8 M urea and 6 M guanidine hydrochloride
  • Avoid using oxidising agents and dextranase
Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
These groups are attached to glucose units in the matrix by stable ether linkages.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty