Would you like to log in again?
CM Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
Sephadex™
17-0210-01 17-0210-01 17-0210-02 17-0210-02
95055-936EA 572.85 USD InStock
95055-936 CA95016-808L CA95055-936L 95016-808
CM Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix
  • Weak cation exchanger suitable batch techniques
  • High binding capacities
  • Avoid using oxidising agents and dextranase
CM Sephadex A-25 is a weak cation exchanger based on the well documented and well proven Sephadex base matrix. Sephadex ion exchangers are produced by introducing functional groups onto the cross-linked dextran matrix. These groups are attached to glucose units in the matrix by stable ether linkages. Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available. Different types of ion exchangers (DEAE, QAE and CM) and two different porosities (A-25/C-25 and A-50/C-50) are available.
These groups are attached to glucose units in the matrix by stable ether linkages.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty