Would you like to log in again?
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sephadex™
17-0230-01 17-0230-01
95055-940EA 577.39 USD InStock
CA95055-940L 95055-940
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
SP Sephadex™ C-25 is a strong cation exchanger.
The ion exchange group is a sulphopropyl group which remains charged and maintains consistently high capacity over the entire working range of pH 4 to 13.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty