Would you like to log in again?
HiTrap Columns, Protein G HP, Cytiva
Supplier: Cytiva
HiTrap™
17-0405-03 17-0405-01 17-0404-01 17-0404-03 17-0404-01 17-0405-01 29-0485-81 17-0404-03 29-0485-81 17-0405-03
89501-370EA 252.7 USD InStock
89501-370 95055-848 95055-946 CA95055-946L CA95055-944L 95055-944 CA95055-848L 95056-034 CA95056-034L CA11033-010
HiTrap Columns, Protein G HP, Cytiva
Chromatography Columns
HiTrap™ Protein G HP affinity columns are pre-packed with 1 or 5 ml of Protein G Sepharose™ high performance for purification and isolation of monoclonal and polyclonal IgG from ascites, serum and cell culture supernatants.
  • Can be operated with a syringe, peristaltic pump or liquid chromatography system such as ÄKTAdesign™ or FPLC™ system
  • Polypropylene column material which is biocompatible and non interactive with biomolecules
  • Top and bottom frits are manufactured from porous polyethylene
Delivery information: Delivered with a stopper on the inlet and a snap-off end on the outlet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty