Would you like to log in again?
HiTrap Columns, Blue HP, Cytiva
Supplier: Cytiva
HiTrap™
17-0413-01 17-0412-01 17-0413-01 17-0412-01
95056-040PK 267.22 USD InStock
95056-040 CA95055-950L CA95056-040L 95055-950
HiTrap Columns, Blue HP, Cytiva
Chromatography Columns
HiTrap™ Blue HP is prepacked with blue Sepharose™ HP for capture of a wide range of biomolecules such as albumin, interferon, a broad range of nucleotide-dependent enzymes, α2-macroglobulin, coagulation factors and related proteins.
  • Can be used for depletion of albumin from serum
  • The ligand, Cibacron Blue F3G-A, is covalently attached to the highly cross-linked agarose medium and is stable over a pH range of 4 to 12 for use and 3 to 13 for cleaning
  • Can be operated with a syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC System
Blue Sepharose High Performance is a group specific adsorbent with affinity for a wide variety of enzymes. The dye ligand, Cibacron Blue F3G-A, is covalently attached to the matrix via the triazine part of the dye molecule. Some proteins interact biospecifically with the dye due to its structural similarity with nucleotide cofactors, while others, such as albumin and interferon, bind in a less specific manner by electrostatic and/or hydrophobic interactions with the aromatic anionic ligand.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty