Would you like to log in again?
HiTrap Columns, NHS-Activated HP, Cytiva
Supplier: Cytiva
HiTrap™
17-0716-01 17-0717-01 17-0717-01 17-0716-01
95055-954EA 225.85 USD InStock
CA95055-952L CA95055-954L 95055-954 95055-952
HiTrap Columns, NHS-Activated HP, Cytiva
Chromatography Columns
HiTrap NHS-activated HP prepacked chromatography columns allow you to design an affinity column suitable for your purification needs.
  • Easy coupling of any ligand containing primary amine groups
  • Packed with NHS-Activated Sepharose High Performance resin
  • Convenient prepacked columns can be used with a syringe, pump, or chromatographic system, such as ÄKTA systems
This NHS-activated resin can be used with any primary amine-containing ligand. The resin reacts with primary amine groups to form stable amide linkages that covalently immobilize antibodies or other proteins for affinity purification. The resin enables simple coupling, carried out at close to neutral pH.
The NHS-activated Sepharose High Performance (HP) resin in these columns is delivered in 100% isopropanol to protect the active groups. It contains ~ 10 μmol NHS groups/ml resin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty