Would you like to log in again?
IMAC Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Sepharose™
17-0920-07 17-0920-07
95055-956EA 1461.99 USD InStock
CA95055-956L 95055-956
IMAC Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
IMAC Sepharose High Performance media is easily charged by the user with the metalion of choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent purification of polyhistidine tagged proteins.
  • For optimizing purification of histidine-tagged proteins when Ni2+ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • 34 μm bead size for high performance purification
  • High binding capacity
  • Stable in commonly used aqueous buffers
  • Avoid using chelating agents, e.g. EDTA, EGTA, citrate
Conveniently charge with your metal of choice. IMAC Sepharose FF is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.g. Cu₂₊, Zn₂₊, Ni₂₊, or Co₂₊); these metal ions will bind to the covalently immobilized chelating ligand on the Sepharose. Small bead size for high performance purification and high binding capacity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty