Would you like to log in again?
HiTrap Columns, Capto DEAE, Cytiva
Supplier: Cytiva
HiTrap™
28-9165-37 28-9165-40 28-9165-40 28-9165-37
95056-246PK 271.59 USD InStock
CA95056-246L 95056-246 95055-980 CA95055-980L
HiTrap Columns, Capto DEAE, Cytiva
Chromatography Columns
HiTrap Capto DEAE is a weak anion exchanger prepacked with BioProcess Capto resins for screening and small-scale protein purifications using ion exchange chromatography (IEX)..
  • High-throughput resin for capture and intermediate purification, excellent choice for process-scale purification
  • The combined properties of high binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
Capto DEAE has a weak diethylaminoethyl anion exchanger coupled to a chemically modified, high-flow agarose matrix. The modified agarose matrix provides greater particle rigidity without compromising pore size, which allows more flexibility in process-scale applications. High dynamic binding capacity is achieved with a dextran surface extender that coats the agarose matrix.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty