Would you like to log in again?
HiPrep Sephacryl S-100 HR, Cytiva
Supplier: Cytiva
HiPrep™
17-1194-01 17-1194-01 17-1165-01 17-1165-01
95055-806EA 1320.63 USD InStock
95055-806 CA95055-806L 95056-000 CA95056-000L
HiPrep Sephacryl S-100 HR, Cytiva
Chromatography Columns
HiPrep Sephacryl S-100 HR are prepacked gel filtration columns for preparative separation of biomolecules from up to 13 ml sample volume.
  • Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Gel filtration columns for low to medium pressure ÄKTA systems.
  • Well-established in industrial production
  • The HiPrep Sephacryl series offers reproducible separations over a wide molecular weight range
  • Convenient, prepacked HiPrep Sephacryl columns speed up protein purification
Sephacryl S-100 is used for separating peptides and small proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty