Would you like to log in again?
Glutathione Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sepharose™
17-5279-02 17-5279-02 17-5279-01 17-5279-01
95056-016EA 2949.98 USD InStock
CA95056-016L CA95056-014L 95056-016 95056-014
Glutathione Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Glutathione Sepharose High Performance is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins.
  • 34 µm bead size for improved efficiency and resolution
  • Fast, one-step purification
  • Mild conditions during the purification process preserves protein antigenicity and function
GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. Fast, one-step purification of GST-tagged proteins and other glutathione S-trasferase or glutathione-dependent proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty