Would you like to log in again?
GSTrap HP Columns, Cytiva
Supplier: Cytiva
GSTrap™
17-5281-01 17-5282-01 17-5282-02 17-5282-02 17-5282-01 17-5281-01
95056-020PK 2149.07 USD InStock
CA95056-212L 95056-020 CA95056-018L CA95056-020L 95056-018 95056-212
GSTrap HP Columns, Cytiva
Chromatography Columns
GSTrap™ HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins.
  • High resolution purification of GST-tagged proteins
  • Mild elution conditions preserving protein antigenicity and function
  • Fast, one-step purification
  • Easy to use with syringe, pump or a chromatography system
GSTrap™ HP 1 and 5 ml columns are prepacked with Glutathione Sepharose High Performance resin, which provides high-resolution purifications, thus a more concentrated sample. GST-tagged proteins are purified from pretreated bacterial lysates by affinity chromatography using immobilized glutathione, such as Glutathione Sepharose High Performance. GST-tagged proteins are bound to the affinity resin, and impurities are removed by washing with binding buffer
GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty