Would you like to log in again?
HisTrap Columns, FF, Cytiva
Supplier: Cytiva
Histrap™
17-5255-01 17-5255-01 17-5319-01 17-5319-01
95056-012PK 851.38 USD InStock
CA95056-012L 95056-012 95056-022 CA95056-022L
HisTrap Columns, FF, Cytiva
Chromatography Columns
HisTrap FF is a ready-to-use column, prepacked with precharged Ni Sepharose 6 Fast Flow for preparative purification of histidine-tagged recombinant proteins by immobilized metal ion affinity chromatography (IMAC).
  • High binding capacity, approx. 40 mg/ml medium
  • Negligible leakage of Ni²⁺
  • Prepacked columns
  • Compatible with a wide range of reducing agents, detergents, denaturants, and additives
This chelating ligand is charged with Ni²⁺ ions, the first-choice metal ion for purifying most histidine-tagged proteins. The negligible leakage of Ni²⁺ ions from the matrix ensures reliable capture of histidine-tagged proteins in repeated IMAC purifications. The high flow rates made possible by the Ni Sepharose 6 Fast Flow matrix make HisTrap FF columns well suited for small scale-up. Capacity can be increased by connecting columns in series. HiTrap IMAC FF is the product of choice when charging the medium with different metal ions for optimization of purification protocols.
Ni Sepharose 6 Fast Flow consists of 90 µm beads of highly cross-linked agarose, to which a chelating ligand has been immobilized. Prepacked columns offer reliable and convenient time-saving purification of histidinetagged recombinant proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty