Would you like to log in again?
Sepharose™ 6B Gel Filtration Base Matrix
Supplier: Cytiva
Sepharose™
17-0110-01 17-0110-01
95056-026EA 1129.3 USD InStock
CA95056-026L 95056-026
Sepharose™ 6B Gel Filtration Base Matrix
Protein Purification Protein Purification Resins
Sepharose™ 6B is an agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
  • 6% agarose gel filtration media
  • Proven base matrix for coupling affinity ligands
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty