Would you like to log in again?
HiTrap Columns, Protein A HP, Cytiva
Supplier: Cytiva
HiTrap™
17-0402-01 17-0402-03 17-0402-03 17-0403-01 17-0403-03 29-0485-76 17-0403-01 17-0402-01 29-0485-76 17-0403-03
95056-032PK 847.17 USD InStock
95056-032 95056-142 CA11033-008 CA95055-846L 95055-846 CA95055-844L 95055-844 89501-368 CA95056-142L CA95056-032L
HiTrap Columns, Protein A HP, Cytiva
Chromatography Columns
HiTrap Protein A HP is a prepacked Protein A Sepharose High Performance column for preparative purification of monoclonal and polyclonal antibodies from ascites, serum, and cell culture supernatants.
  • High binding capacitiy of 20 mg IgG/ml medium and fast kinetics
  • 34 µm bead size for high-performance, high-resolution purifications
  • Designed for use with syringe, pump, or chromatography system such as ÄKTA design or FPLC System
  • Stable over pH 3 to 9 for use, and 2 to 10 for cleaning
  • Robust, easy-to-use method
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty