Would you like to log in again?
HiTrap Columns, IMAC HP, Cytiva
Supplier: Cytiva
HiTrap™
17-0920-05 17-0920-03 17-0920-03 17-0920-05
95056-042PK 277.38 USD InStock
95056-042 95056-152 CA95056-042L CA95056-152L
HiTrap Columns, IMAC HP, Cytiva
Chromatography Columns
HiTrap IMAC HP is prepacked with IMAC Sepharose High Performance. The resin is charged with the metalion of your choice for Immobilized Metal ion Affinity Chromatography (IMAC) and subsequent high-resolution purification of polyhistidine tagged proteins.
  • For optimizing purification of histidine-tagged proteins when Ni2+ is not the best choice of metal ion
  • Conveniently charge with your metal of choice
  • Small bead size for high-performance purification
  • High binding capacity
IMAC Sepharose HP is supplied free of metal ions. It is charged by the user with the transition metal ion of choice (e.g. Cu²⁺, Zn²⁺, Ni²⁺, or Co²⁺); these metal ions will bind to the covalently immobilized chelating ligand on the Sepharose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty