Would you like to log in again?
GSTrap FF Columns, Cytiva
Supplier: Cytiva
GSTrap™
17513001 17-5131-02 17-5131-01 17-5131-02 17-5130-02 17-5130-02 17-5131-01 17-5130-01
95056-096PK 1936.77 USD InStock
95056-096 95056-092 95056-094 CA95056-092L CA95056-094L CA95056-096L CA95055-894L 95055-894
GSTrap FF Columns, Cytiva
Chromatography Columns
GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.
  • Fast and simple one-step purification of GST-tagged proteins
  • Mild elution conditions preserving protein antigenicity and function
  • Easy scale-up by connecting the columns in series
  • Sepharose 4 Fast Flow provides good flow properties
GST-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrap FF.GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione.
The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods.
The resin, Glutathione Sepharose 4 Fast Flow, is also available as lab packages and is an excellent choice for scale-up.The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty