Would you like to log in again?
HiTrap Columns, DEAE Sepharose FF, Cytiva
Supplier: Cytiva
HiTrap™
17-5055-01 17-5154-01 17-5154-01 17-5055-01
95056-084PK 195.88 USD InStock
95056-084 CA95056-084L 95056-100 CA95056-100L
HiTrap Columns, DEAE Sepharose FF, Cytiva
Chromatography Columns
HiTrap DEAE FF is prepacked with DEAE Sepharose Fast Flow, and is a weak anion exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
  • Convenient and affordable for fast, easy ion exchange separations either alone or connected in series
  • The industry standard for ion exchange chromatography
  • High flow rates and good scale-up potential
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
  • Predictable scale-up
The active end of the charged group is the same for DEAE Sepharose Fast Flow and ANX Sepharose Fast Flow (high sub), the difference is the length of the carbon chain of the charged group. DEAE Sepharose Fast Flow has a diethylaminoethyl-group bound to the agarose whilst ANX Sepharose 4 Fast Flow (high sub) has a diethylaminopropyl group attached.
Q, SP, DEAE, and CM Sepharose Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose 4 Fast Flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity, which is particularly useful for the purification of high molecular mass proteins.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty