Would you like to log in again?
HiTrap Columns, Q Sepharose FF, Cytiva
Supplier: Cytiva
HiTrap™
17-5156-01 17-5156-01 17-5053-01 17-5053-01
95056-080PK 199.65 USD InStock
CA95056-104L CA95056-080L 95056-080 95056-104
HiTrap Columns, Q Sepharose FF, Cytiva
Chromatography Columns
HiTrap Q FF is prepacked with Q Sepharose Fast Flow, and is a strong anion exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
  • Convenient and affordable for fast, easy ion exchange separations either alone or connected in series
  • The industry standard for ion exchange chromatography
  • High flow rates and good scale-up potential
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
  • Predictable scale-up
Composed of crosslinked 6% agarose beads with quaternary amine strong anion exchange groups, this resin has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols. The resin’s 90 µm bead size yields rapid flow rates, making it simple to scale up the purification.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty