Would you like to log in again?
HiTrap Columns, SP Sepharose FF, Cytiva
Supplier: Cytiva
HiTrap™
17-5157-01 17-5054-01 17-5054-01 17-5157-01
95056-082PK 192.04 USD InStock
CA95056-106L CA95056-082L 95056-082 95056-106
HiTrap Columns, SP Sepharose FF, Cytiva
Chromatography Columns
HiTrap SP FF is prepacked with SP Sepharose Fast Flow, and is a strong cation exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.
  • Convenient and affordable for fast, easy ion exchange separations
  • The industry standard for ion exchange chromatography
  • High flow rates and good scale-up potential
  • Use a weak ion exchanger if the selectivity of a strong ion exchanger is insufficient
  • Predictable scale-up
Q, SP, DEAE, and CM Sepharose™ Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty